Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The protein encoded by this gene is a member of the keratin gene family. As a type II hair keratin, it is a basic protein which heterodimerizes with type I keratins to form hair and nails. The type II hair keratins are clustered in a region of chromosome 12q13 and are grouped into two distinct subfamilies based on structure similarity. One subfamily, consisting of KRTHB1, KRTHB3, and KRTHB6, is highly related. The other less-related subfamily includes KRTHB2, KRTHB4, and KRTHB5. All hair keratins are expressed in the hair follicle; this hair keratin, as well as KRTHB3 and KRTHB6, is found primarily in the hair cortex. Mutations in this gene and KRTHB6 have been observed in patients with a rare dominant hair disease, monilethrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ghHb1; ghHKb1; Hair keratin K2.9; hard keratin, type II, 1; K81; keratin 81, type II; keratin complex 2, basic, gene 19; Keratin, hair, basic, 1; Keratin, type II cuticular Hb1; Keratin-81; krt81; Metastatic lymph node 137 gene protein; MLN 137; Type II hair keratin Hb1; Type-II keratin Kb21
Gene Aliases: AA589625; ghHkb1; Hb-1; HB1; hHAKB2-1; K81; Krt2-19; KRT81; KRTHB1; MLN137; MNLIX2
UniProt ID: (Human) Q14533, (Mouse) Q9ERE2
Entrez Gene ID: (Human) 3887, (Mouse) 64818
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support