Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077057.2 | NP_001070525.1 | 2 | 649 | 142 | |
GenBank mRNA | BC109836.1 | - | 2 | 650 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |