Search
Search

tlx1| Gene Symbol: tlx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170765.1 | NP_739571.1 | 1 - 2 | 570 | 82 | |
| GenBank mRNA | AF398517.1 | - | 1 - 2 | 570 | 82 | |
| BC162115.1 | - | 1 - 2 | 507 | 82 | ||
| BC162486.1 | - | 1 - 2 | 507 | 82 | ||
Set Membership: |
|