Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012917.1 | NP_001012935.1 | 6 - 7 | 1294 | 84 | |
XM_015289441.2 | XP_015144927.2 | 6 - 7 | 1475 | 84 | ||
XM_015289442.2 | XP_015144928.2 | 5 - 6 | 1325 | 84 | ||
XM_015289443.2 | XP_015144929.2 | 5 - 6 | 1211 | 84 | ||
XM_015289444.2 | XP_015144930.2 | 5 - 6 | 1339 | 84 | ||
XM_015289445.2 | XP_015144931.2 | 5 - 6 | 1262 | 84 | ||
XM_025152191.1 | XP_025007959.1 | 6 - 7 | 1754 | 84 | ||
GenBank mRNA | AJ851593.1 | - | 6 - 7 | 1294 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |