Search
Search

SCRN1| Gene Symbol: SCRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549892.2 | XP_005549949.1 | 2 - 3 | 332 | 100 | |
| XM_005549893.2 | XP_005549950.1 | 2 - 3 | 920 | 100 | ||
| XM_015447582.1 | XP_015303068.1 | 2 - 3 | 177 | 100 | ||
Set Membership: |
|