Search
Search

Dtnbp1| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025772.4 | NP_080048.2 | 3 - 4 | 289 | 79 | |
| GenBank mRNA | AJ404859.1 | - | 3 - 4 | 288 | 79 | |
| AK150306.1 | - | 3 - 4 | 285 | 79 | ||
| AK159656.1 | - | 3 - 4 | 414 | 79 | ||
| BC018350.1 | - | 3 - 4 | 280 | 4195234 | 79 | |
| BC048682.1 | - | 3 - 4 | 334 | 79 | ||
| BC058574.1 | - | 3 - 4 | 262 | 6487720 | 79 | |
Set Membership: |
|