Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100063.2 | XP_014955549.2 | 1 - 2 | 229 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |