Search
Search

UFD1| Gene Symbol: UFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036307.2 | NP_001031384.1 | 7 - 8 | 1159 | 85 | |
| NM_001084464.1 | NP_001077933.1 | 8 - 9 | 951 | 85 | ||
| NM_001335738.1 | NP_001323742.1 | 8 - 9 | 1015 | 85 | ||
| NM_001335739.1 | NP_001323741.1 | 8 - 9 | 934 | 85 | ||
| NM_127699.5 | NP_565504.1 | 8 - 9 | 715 | 85 | ||
| GenBank mRNA | AK226526.1 | - | 8 - 9 | 716 | 85 | |
| AK317375.1 | - | 8 - 9 | 952 | 85 | ||
| BT033134.1 | - | 6 - 7 | 613 | 85 | ||
Set Membership: |
|