Search
Search

NAS1| Gene Symbol: NAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120577.4 | NP_196114.1 | 1 | 1100 | 103 | |
| GenBank mRNA | AY072364.1 | - | 1 | 921 | 103 | |
| BT000392.1 | - | 1 | 899 | 103 | ||
Set Membership: |
|