Search
Search

AT4G17950| Gene Symbol: AT4G17950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117905.3 | NP_567546.1 | 2 - 3 | 922 | 79 | |
| GenBank mRNA | AY054610.1 | - | 2 - 3 | 922 | 79 | |
| AY081495.1 | - | 2 - 3 | 697 | 79 | ||
Set Membership: |
|