Search
Search

bZIP| Gene Symbol: bZIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105562.3 | NP_177054.1 | 1 | 502 | 69 | |
| GenBank mRNA | AB493528.1 | - | 1 | 361 | 69 | |
| AF400621.1 | - | 1 | 361 | 69 | ||
| BT024817.1 | - | 1 | 361 | 69 | ||
Set Membership: |
|