Search
Search

AT2G47780| Gene Symbol: AT2G47780 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130345.5 | NP_182299.1 | 1 - 2 | 365 | 77 | |
| GenBank mRNA | BT000468.1 | - | 1 - 2 | 356 | 77 | |
| BT002146.1 | - | 1 - 2 | 329 | 77 | ||
Set Membership: |
|