Search
Search

ABCA11| Gene Symbol: ABCA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345487.1 | NP_001318858.1 | 7 - 8 | 1802 | 127 | |
| NM_001345488.1 | NP_001332171.1 | 7 - 8 | 1802 | 127 | ||
Set Membership: |
|