Search
Search

ALDH2B7| Gene Symbol: ALDH2B7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332622.1 | NP_001321290.1 | 4 - 5 | 581 | 75 | |
| NM_102228.4 | NP_564204.1 | 4 - 5 | 581 | 75 | ||
| GenBank mRNA | AY035139.1 | - | 4 - 5 | 573 | 75 | |
| AY113912.1 | - | 4 - 5 | 499 | 75 | ||
Set Membership: |
|