Search
Search

RING1A| Gene Symbol: RING1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161288.1 | NP_001154760.1 | 7 - 8 | 780 | 149 | |
| NM_001344539.1 | NP_001331587.1 | 4 - 5 | 346 | 149 | ||
| NM_123795.5 | NP_199241.2 | 5 - 6 | 820 | 149 | ||
Set Membership: |
|