Search
Search

AT4G25310| Gene Symbol: AT4G25310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341727.1 | NP_001329605.1 | 4 - 5 | 813 | 148 | |
| NM_118663.3 | NP_194261.1 | 3 - 4 | 897 | 148 | ||
| GenBank mRNA | BT011229.1 | - | 3 - 4 | 897 | 148 | |
| BT012157.1 | - | 3 - 4 | 808 | 148 | ||
Set Membership: |
|