Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024524.3 | NP_001019695.2 | 3 - 4 | 583 | 74 | |
GenBank mRNA | BC126551.1 | - | 3 - 4 | 584 | 74 | |
BT021593.1 | - | 3 - 4 | 590 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |