Search
Search

pes-5| Gene Symbol: pes-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267388.2 | NP_001254317.1 | 2 - 3 | 1016 | 98 | |
| NM_001373797.2 | NP_001359621.1 | 1 - 2 | 914 | 98 | ||
| NM_001381619.1 | NP_001366978.1 | 1 - 2 | 878 | 98 | ||
Set Membership: |
|