Search
Search
| Gene Symbol: HPRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003357.2 | NP_001003357.2 | 8 - 9 | 756 | 129 | |
| NM_001313818.1 | NP_001300747.1 | 7 - 8 | 649 | 129 | ||
| GenBank mRNA | AY283372.1 | - | 8 - 9 | 607 | 129 | |
Set Membership: |
|