Search
Search

MIR8870| Gene Symbol: MIR8870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128823.1 | 1 | 68 | 107 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WSC domain containing 1 | WSCD1 | Sense | Chr. 5 - 30992773 - 31023526 on NCBI Build CanFam3.1 | ||
| Downstream | NLR family pyrin domain containing 1 | NLRP1 | 261.8kb | Anti-sense | Chr. 5 - 31264554 - 31299538 on NCBI Build CanFam3.1 |
Set Membership: |
|