Search
Search

Creld2| Gene Symbol: Creld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244136.1 | NP_001231065.1 | 3 - 4 | 503 | 57 | |
| GenBank mRNA | JI867916.1 | - | 1 - 2 | 92 | 57 | |
| JP042630.1 | - | 3 - 4 | 268 | 57 | ||
| U48852.1 | - | 3 - 4 | 503 | 57 | ||
Set Membership: |
|