Search
Search

smal| Gene Symbol: smal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103627.4 | NP_001097097.1 | 5 - 6 | 1311 | 58 | |
| NM_001273195.1 | NP_001260124.1 | 5 - 6 | 1311 | 58 | ||
| NM_164688.2 | NP_723165.2 | 5 - 6 | 1311 | 58 | ||
Set Membership: |
|