Search
Search

Etf-QO| Gene Symbol: Etf-QO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259302.1 | NP_001246231.1 | 5 - 6 | 1581 | 78 | |
| NM_136692.1 | NP_610536.1 | 4 - 5 | 1638 | 78 | ||
| GenBank mRNA | AY113586.1 | - | 4 - 5 | 1638 | 78 | |
Set Membership: |
|