Search
Search

CG12096| Gene Symbol: CG12096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298261.1 | NP_001285190.1 | 1 - 2 | 1216 | 90 | |
| NM_001298262.1 | NP_001285191.1 | 1 - 2 | 1216 | 90 | ||
| NM_132634.4 | NP_572862.2 | 1 - 2 | 1426 | 90 | ||
| GenBank mRNA | BT133216.1 | - | 1 - 2 | 1156 | 90 | |
Set Membership: |
|