Search
Search

Men| Gene Symbol: Men | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275580.1 | NP_001262509.1 | 1 - 2 | 395 | 54 | |
| NM_080141.4 | NP_524880.2 | 2 - 3 | 937 | 54 | ||
| NM_169479.2 | NP_731739.1 | 2 - 3 | 798 | 54 | ||
| GenBank mRNA | AY118540.1 | - | 2 - 3 | 1040 | 54 | |
| BT100054.1 | - | 2 - 3 | 785 | 54 | ||
| DQ148978.1 | - | 1 - 2 | 137 | 54 | ||
| DQ148979.1 | - | 1 - 2 | 137 | 54 | ||
| DQ148980.1 | - | 1 - 2 | 137 | 54 | ||
| DQ148981.1 | - | 1 - 2 | 137 | 54 | ||
| DQ148982.1 | - | 1 - 2 | 137 | 54 | ||
| DQ148983.1 | - | 1 - 2 | 137 | 54 | ||
| DQ148988.1 | - | 1 - 2 | 137 | 54 | ||
| DQ148991.1 | - | 1 - 2 | 137 | 54 | ||
| DQ148992.1 | - | 1 - 2 | 137 | 54 | ||
| DQ148994.1 | - | 1 - 2 | 137 | 54 | ||
| DQ148997.1 | - | 1 - 2 | 137 | 54 | ||
| DQ148998.1 | - | 1 - 2 | 137 | 54 | ||
| DQ149000.1 | - | 1 - 2 | 137 | 54 | ||
| DQ149001.1 | - | 1 - 2 | 137 | 54 | ||
| DQ149002.1 | - | 1 - 2 | 137 | 54 | ||
| DQ149003.1 | - | 1 - 2 | 137 | 54 | ||
| DQ149004.1 | - | 1 - 2 | 137 | 54 | ||
| DQ149005.1 | - | 1 - 2 | 137 | 54 | ||
| DQ149006.1 | - | 1 - 2 | 137 | 54 | ||
| DQ149009.1 | - | 1 - 2 | 137 | 54 | ||
| JQ690827.1 | - | 1 - 2 | 173 | 54 | ||
| JQ690831.1 | - | 1 - 2 | 173 | 54 | ||
| JQ690840.1 | - | 1 - 2 | 149 | 54 | ||
| JQ690841.1 | - | 1 - 2 | 173 | 54 | ||
| JQ690842.1 | - | 1 - 2 | 173 | 54 | ||
| JQ690843.1 | - | 1 - 2 | 173 | 54 | ||
| JQ690854.1 | - | 1 - 2 | 173 | 54 | ||
| JQ690856.1 | - | 1 - 2 | 173 | 54 | ||
| JQ690859.1 | - | 1 - 2 | 173 | 54 | ||
Set Membership: |
|