Search
Search

l(2)efl| Gene Symbol: l(2)efl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274227.1 | NP_001261156.1 | 2 - 3 | 460 | 131 | |
| NM_079103.3 | NP_523827.1 | 2 - 3 | 457 | 131 | ||
| GenBank mRNA | AY047516.1 | - | 2 - 3 | 435 | 131 | |
Set Membership: |
|