Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080059.1 | NP_001073528.1 | 3 - 4 | 296 | 152 | |
GenBank mRNA | EF010535.1 | - | 3 - 4 | 296 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |