Search
Search

MIR551A| Gene Symbol: MIR551A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032807.1 | 1 | 30 | 56 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | multiple EGF like domains 6 | MEGF6 | Sense | Chr. 2 - 46718631 - 46820967 on NCBI Build EquCab3.0 | ||
| Downstream | Rho guanine nucleotide exchange factor 16 | ARHGEF16 | 69.0kb | Anti-sense | Chr. 2 - 46826907 - 46850802 on NCBI Build EquCab3.0 |
Set Membership: |
|