Search
Search

ATP6AP2| Gene Symbol: ATP6AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309406.1 | NP_001296335.1 | 3 - 4 | 398 | 60 | |
| GenBank mRNA | CD536448.1 | - | 3 - 4 | 398 | 60 | |
| JL617525.1 | - | 3 - 4 | 334 | 60 | ||
Set Membership: |
|