Search
Search

LYPD4| Gene Symbol: LYPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596295.3 | XP_005596352.1 | 1173 | 104 | ||
| XM_005596296.3 | XP_005596353.1 | 1211 | 104 | |||
| XM_023651444.1 | XP_023507212.1 | 345 | 104 | |||
| XM_023651445.1 | XP_023507213.1 | 1077 | 104 | |||
| XM_023651446.1 | XP_023507214.1 | 325 | 104 | |||
| XM_023651447.1 | XP_023507215.1 | 313 | 104 | |||
Set Membership: |
|