Search
Search

C18H2orf76| Gene Symbol: C18H2orf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005601445.3 | XP_005601502.1 | 423 | 100 | ||
| XM_023622778.1 | XP_023478546.1 | 632 | 100 | |||
| XM_023622779.1 | XP_023478547.1 | 620 | 100 | |||
| XM_023622780.1 | XP_023478548.1 | 291 | 100 | |||
| XM_023622781.1 | XP_023478549.1 | 424 | 100 | |||
| XM_023622782.1 | XP_023478550.1 | 535 | 100 | |||