Search
Search

FTL| Gene Symbol: FTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204383.1 | NP_989714.1 | 1 - 2 | 170 | 61 | |
| GenBank mRNA | AF447376.1 | - | 1 - 2 | 170 | 61 | |
| BX935096.2 | - | 1 - 2 | 244 | 61 | ||
Set Membership: |
|