Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000232014.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000406870.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000419510.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000430339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000430339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000430339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000430339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000430339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000430339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000438077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000438077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000438077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000438077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000438077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000450123.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000450123.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000450123.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000450123.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000450123.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000450123.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000450123.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000450123.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000450123.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000450123.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000470319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000470319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000470319.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000479110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000479110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000479110.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000480458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000480458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000480458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000480458.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000496823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000496823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000496823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000496823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000496823.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
ENST00000621333.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001130845.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001134738.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013471.hg.1 |
NM_001706.4 |
Refseq |