| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019596.hg.1 |
ENST00000374037.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019596.hg.1 |
ENST00000544425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019596.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019596.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06019596.hg.1 |
TC06001658.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600179494.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179494.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179494.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179494.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179494.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179494.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600179494.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179494.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600179494.hg.1 |
TC0600011590.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000374037.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
ENST00000544425.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_001252294.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011590.hg.1 |
NM_012391.2 |
Refseq |