| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000586287.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000586287.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000586287.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589614.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589614.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589614.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589614.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000589882.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020461.hg.1 |
ENST00000244709.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020461.hg.1 |
ENST00000334475.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020461.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020461.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020461.hg.1 |
TC06001717.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020462.hg.1 |
ENST00000244709.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020462.hg.1 |
ENST00000334475.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020462.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020462.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020462.hg.1 |
TC06001717.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020464.hg.1 |
ENST00000244709.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020464.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020464.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020464.hg.1 |
TC06001717.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020460.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020460.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06020460.hg.1 |
TC06001717.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000586287.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000586287.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000586287.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589614.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589614.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589614.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589614.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000589882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011777.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180877.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180877.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180877.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180877.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180877.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180877.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180877.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180877.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180877.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180877.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180877.hg.1 |
TC0600011777.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
ENST00000589614.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
ENST00000589882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
ENST00000589614.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
ENST00000589882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
ENST00000591620.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
NM_001242589.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180882.hg.1 |
TC0600011777.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600180874.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180874.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180874.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180874.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180874.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180874.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180874.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180874.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180874.hg.1 |
TC0600011777.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600180873.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180873.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180873.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180873.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180873.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180873.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180873.hg.1 |
TC0600011777.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600180878.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180878.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180878.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180878.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180878.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180878.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180878.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180878.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180878.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180878.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180878.hg.1 |
TC0600011777.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600180876.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180876.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180876.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180876.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180876.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180876.hg.1 |
ENST00000244709.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180876.hg.1 |
ENST00000334475.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180876.hg.1 |
ENST00000589695.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600180876.hg.1 |
NM_001242590.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180876.hg.1 |
NM_018643.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600180876.hg.1 |
TC0600011777.hg.1 |
Clariom D TC |