| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021019.hg.1 |
TC01001339.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021019.hg.1 |
ENST00000368169.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021019.hg.1 |
ENST00000368170.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021019.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021022.hg.1 |
TC01001339.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021022.hg.1 |
ENST00000368169.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021022.hg.1 |
ENST00000368170.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021022.hg.1 |
ENST00000454192.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021022.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021021.hg.1 |
TC01001339.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021021.hg.1 |
ENST00000368169.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021021.hg.1 |
ENST00000368170.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021021.hg.1 |
ENST00000454192.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021021.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000443761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000443761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000443761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
ENST00000443761.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021023.hg.1 |
TC01001339.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021023.hg.1 |
ENST00000368169.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021023.hg.1 |
ENST00000368170.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021023.hg.1 |
ENST00000454192.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021023.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021018.hg.1 |
TC01001339.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021018.hg.1 |
ENST00000368169.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021018.hg.1 |
ENST00000368170.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01021018.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175040.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175040.hg.1 |
ENST00000443761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175040.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175040.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175040.hg.1 |
ENST00000443761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175040.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175040.hg.1 |
TC0100010230.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100175038.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175038.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175038.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175038.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175038.hg.1 |
TC0100010230.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100175035.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175035.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175035.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175035.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175035.hg.1 |
TC0100010230.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100175039.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175039.hg.1 |
ENST00000443761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175039.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175039.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175039.hg.1 |
ENST00000443761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175039.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175039.hg.1 |
TC0100010230.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000443761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000443761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000443761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
ENST00000443761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100010230.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175037.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175037.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175037.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175037.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175037.hg.1 |
TC0100010230.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100175036.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175036.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175036.hg.1 |
ENST00000368170.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100175036.hg.1 |
NM_001765.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100175036.hg.1 |
TC0100010230.hg.1 |
Clariom D TC |