| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012658.hg.1 |
ENST00000380232.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012658.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012658.hg.1 |
TC09000946.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012657.hg.1 |
ENST00000380232.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012657.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012657.hg.1 |
TC09000946.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900166293.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166293.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166293.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166293.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166293.hg.1 |
TC0900009688.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900166295.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166295.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166295.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166295.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166295.hg.1 |
TC0900009688.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009688.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166294.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166294.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166294.hg.1 |
ENST00000380232.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166294.hg.1 |
NM_002176.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166294.hg.1 |
TC0900009688.hg.1 |
Clariom D TC |