| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02021088.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02021088.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02021088.hg.1 |
TC02001351.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200178253.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200178253.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200178253.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200178253.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200178253.hg.1 |
TC0200010911.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000281830.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
ENST00000488477.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010911.hg.1 |
NM_080671.3 |
Refseq |