| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008183.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22010166.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22010166.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22010166.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22010166.hg.1 |
TC22000567.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22010167.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22010167.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22010167.hg.1 |
TC22000567.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008183.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200158633.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200158633.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200158633.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200158633.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200158633.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200158633.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200158633.hg.1 |
TC2200008183.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200158634.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200158634.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200158634.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200158634.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200158634.hg.1 |
TC2200008183.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200158632.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200158632.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200158632.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200158632.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200158632.hg.1 |
ENST00000398465.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200158632.hg.1 |
NM_013378.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200158632.hg.1 |
TC2200008183.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200158635.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200158635.hg.1 |
ENST00000248948.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200158635.hg.1 |
TC2200008183.hg.1 |
Clariom D TC |