Search
Search

SCT| Gene Symbol: SCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021920.4 | NP_068739.1 | 2 - 3 | 214 | 68 | |
| GenBank mRNA | HY019819.1 | - | 2 - 3 | 273 | 68 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07595488 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27989891 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07595489 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P104594 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P104594 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 20008 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20213 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 41859 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P104594 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P104594 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595488 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27989891 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27989891 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07595488 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P104594 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07595492 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07595491 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07595489 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07595493 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07595498 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07595496 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07595488 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16406607 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P07595498 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16406621 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16406619 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16406610 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P07595488 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P07595497 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16406620 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16406622 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16406608 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P07595489 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16406606 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16406609 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P104594 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595484 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406608 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595465 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406556 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595488 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406555 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595452 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595478 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595467 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406594 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406603 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406573 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595491 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595458 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406607 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595459 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595496 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406622 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595482 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595463 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406619 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595481 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406592 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595483 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595479 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595495 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406576 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595492 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595457 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406588 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595489 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595493 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595461 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406575 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595498 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406565 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16406558 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595471 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595454 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07595472 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_021920_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7945500 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7945501 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7945502 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7945499 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7945498 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 221198_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11742661_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11742661_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015674.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015674.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015674.hg.1 | TC11001243.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015676.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015676.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015676.hg.1 | TC11001243.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015675.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015675.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015675.hg.1 | TC11001243.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009687.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009687.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009687.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009687.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009687.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009687.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009687.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009687.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009687.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015677.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015677.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015677.hg.1 | TC11001243.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100009687.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009687.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009687.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009687.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009687.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009687.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009687.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009687.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009687.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100173032.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100173032.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100173032.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100173032.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100173032.hg.1 | TC1100009687.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100173031.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100173031.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100173031.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100173031.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100173031.hg.1 | TC1100009687.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100173030.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100173030.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100173030.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100173030.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100173030.hg.1 | TC1100009687.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100173033.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100173033.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100173033.hg.1 | ENST00000176195.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100173033.hg.1 | NM_021920.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100173033.hg.1 | TC1100009687.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 221198_at | - | - |
| Human Genome U133 Plus 2.0 Array | 221198_PM_at | - | - |
| Human Genome U133A 2.0 Array | 221198_PM_at | - | - |
| Human Genome U133A 2.0 Array | 221198_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_021920_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw577537 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221198_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221198_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221198_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221198_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221198_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221198_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221198_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221198_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221198_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221198_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221198_at8 | - | - |
| Human Genome U133 Set | 221198_at | - | - |
| Human Genome U133 Set | 221198_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044724 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044724 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 178364 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1705763 | NM_021920.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1705763 | NM_021920.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1705763 | NM_021920.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1705763 | NM_021920.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1705763 | NM_021920.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_12545379-S | NM_021920.2 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_27664-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0004570170 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0004570170 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1705763 | NM_021920.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1705763 | NM_021920.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 6040224 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_27664 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1705763 | NM_021920.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1705763 | NM_021920.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1705763 | NM_021920.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1705763 | NM_021920.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1705763 | NM_021920.2 | human_transcriptome |