| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014033.hg.1 |
ENST00000300961.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014033.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014033.hg.1 |
TC19001025.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900203499.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203499.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900203499.hg.1 |
TC1900011838.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900203499.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900203499.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
ENST00000300961.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011838.hg.1 |
NM_144616.3 |
Refseq |