| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758028 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897883 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897860 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897848 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897849 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758006 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897876 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758060 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897806 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897807 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897839 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897885 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897865 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897808 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758034 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897840 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897856 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897823 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758043 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897874 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897854 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897875 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758048 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02757998 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758029 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758067 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758044 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758045 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897813 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758069 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897817 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758061 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758037 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897818 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897893 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897833 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897809 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15510092 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758049 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897845 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758007 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897890 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758070 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897888 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897820 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758009 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15510099 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758018 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897841 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02757992 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897862 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897810 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897877 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897896 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897822 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02757988 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758023 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897863 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897870 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15510101 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15510093 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897815 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758041 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758055 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897821 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758010 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897805 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897843 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758004 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758059 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22897826 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02758030 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
105915 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
529434 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
215452 |
AB209877.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
215452 |
AK290177.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
215452 |
AK291705.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
215452 |
AK293027.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
215452 |
AK315812.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
215452 |
AL834279.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
215452 |
BC028362.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
215452 |
BC041121.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
215452 |
M69238.1 |
genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
215452 |
NM_001668.2 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
215452 |
NM_178427.1 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
134769 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
221172 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
129374 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
62781 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
24705 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
15163 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
8073 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
418394 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
90090 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
321833 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
30642 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
321453 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
169465 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
121648 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
7839 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
419930 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
57568 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
339481 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
240254 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
72476 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
27065 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
34999 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
176973 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
267814 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3333762 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_1442 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_HS_44k_1443 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
13145 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_18_P23309325 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P31160020 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P31160044 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P31159965 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P31160075 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P16495341 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3333762 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3333762 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897826 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897805 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897815 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897818 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02758010 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02758006 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897822 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897821 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897823 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02758007 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897808 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897807 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897813 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897809 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897833 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15510093 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02757992 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897820 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02758012 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897806 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897817 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02757998 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897810 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897809 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897808 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02757992 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897820 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897806 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02758007 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897805 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897822 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02758012 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897823 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897833 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02757998 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15510093 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897807 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897821 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897813 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02758006 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897826 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897818 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897815 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02758010 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897810 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P22897817 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P16495338 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P31160085 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P31159965 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P16495318 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P16495325 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P16495341 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P14492748 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758026 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758066 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02757978 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758015 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758055 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02757997 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758058 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758000 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02757988 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02757986 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02757991 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15510089 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758047 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758010 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758025 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758007 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758006 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02757976 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02757985 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758048 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758019 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758033 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758063 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758035 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758105 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758070 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15510091 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758022 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758042 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02757974 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758060 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758057 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758031 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758017 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758011 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15510093 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02757994 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758002 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02757982 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758014 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758037 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758040 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758053 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758072 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758013 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758102 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758068 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02757980 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758044 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02758051 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15510095 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P31159965 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P16495341 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P16495318 |
- |
- |