| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000449812.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000449812.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000449812.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
ENST00000449812.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100008493.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160332.hg.1 |
TC0100008493.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100160345.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160345.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160345.hg.1 |
TC0100008493.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100160329.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160329.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160329.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160329.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160329.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160329.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160329.hg.1 |
TC0100008493.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000449812.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000449812.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000449812.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
ENST00000449812.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100008493.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160344.hg.1 |
ENST00000449812.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160344.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160344.hg.1 |
ENST00000449812.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160344.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160344.hg.1 |
TC0100008493.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160331.hg.1 |
TC0100008493.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
ENST00000424308.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
ENST00000438195.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
ENST00000447329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
NR_110626.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160333.hg.1 |
TC0100008493.hg.1 |
Clariom D TC |