| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000600474.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000600474.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000600474.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000600474.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000600474.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000601354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000601354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000601354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000601354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000601354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
ENST00000601354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004485.hg.1 |
ENST00000252599.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004485.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004485.hg.1 |
TC19000340.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000600474.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000600474.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000600474.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000600474.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000600474.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000601354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000601354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000601354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000601354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000601354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
ENST00000601354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900007331.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900153544.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900153544.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900153544.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900153544.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900153544.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900153544.hg.1 |
ENST00000252599.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900153544.hg.1 |
ENST00000593832.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900153544.hg.1 |
ENST00000597075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900153544.hg.1 |
ENST00000597147.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900153544.hg.1 |
NM_024656.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900153544.hg.1 |
TC1900007331.hg.1 |
Clariom D TC |