| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.1 ST Array |
8065630 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1556721_PM_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1556722_PM_a_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20010191.hg.1 |
ENST00000360785.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20010191.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20010191.hg.1 |
TC20000765.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20010189.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20010189.hg.1 |
TC20000765.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20010190.hg.1 |
ENST00000360785.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20010190.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20010190.hg.1 |
TC20000765.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000161737.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161737.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161737.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161737.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000161737.hg.1 |
TC2000008856.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000161737.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161737.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161737.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161737.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000161733.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161733.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000161733.hg.1 |
TC2000008856.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000161733.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161733.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000161736.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161736.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161736.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161736.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000161736.hg.1 |
TC2000008856.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000161736.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161736.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161736.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161736.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008856.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000161734.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161734.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161734.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161734.hg.1 |
NM_182584.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000161734.hg.1 |
TC2000008856.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000161734.hg.1 |
ENST00000360785.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161734.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161734.hg.1 |
ENST00000608990.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000161734.hg.1 |
NM_182584.3 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
1556722_PM_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1556721_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1556721_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1556722_a_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556722_a_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556721_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556721_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556721_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556722_a_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556722_a_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556721_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556722_a_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556722_a_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556722_a_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556722_a_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556721_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556721_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556721_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556721_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556721_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556721_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556722_a_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556722_a_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556721_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556722_a_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1556722_a_at8 |
- |
- |
| Human Genome U95 Set |
54638_PM_at |
- |
- |
| Human Genome U95 Set |
54638_at |
- |
- |