Search
Search

HIGD2A| Gene Symbol: HIGD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138820.4 | NP_620175.1 | 2 | 562 | 95 | |
| GenBank mRNA | BC007502.2 | - | 2 | 530 | 95 | |
| BQ671009.1 | - | 2 | 496 | 95 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04483629 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04483632 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04483624 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04483631 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04483634 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04483633 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 39423 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 55584 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P304386 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P304386 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 7651 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 10956 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P120140 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P304386 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P304386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483630 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483628 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24664653 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483626 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483630 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24664653 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483628 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483626 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04483624 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P304386 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483633 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483611 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483600 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483622 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483626 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483603 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483605 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483601 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483616 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483631 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483624 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483594 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483598 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483619 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483609 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483608 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483629 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483630 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483612 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483618 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04483615 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04483622 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04483623 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04483628 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04483626 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15767266 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04483624 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15767265 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04483627 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04483621 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P120140 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P304386 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 126292 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P120140 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04483624 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04483631 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04483633 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04483628 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04483630 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P120140 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_138820_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8110168 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8110167 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8110166 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 209328_PM_x_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 209329_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11719580_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11719580_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 209329_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 209329_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 209328_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 209328_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 213833_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 213833_x_at | - | - |
| Human Genome U133A 2.0 Array | 209328_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 209329_x_at | - | - |
| Human Genome U133A 2.0 Array | 209328_x_at | - | - |
| Human Genome U133A 2.0 Array | 209329_PM_x_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_138820_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw597373 | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw567612 | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw570622 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209329_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209329_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209329_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209329_x_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209329_x_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209328_x_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209328_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209329_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209328_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209328_x_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209329_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209328_x_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209328_x_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209328_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209328_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209329_x_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209328_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209329_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209328_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209328_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209329_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 209329_x_at6 | - | - |
| Human Genome U133 Set | 209329_PM_x_at | - | - |
| Human Genome U133 Set | 209329_x_at | - | - |
| Human Genome U133 Set | 209328_x_at | - | - |
| Human Genome U133 Set | 209328_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029204 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029204 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 106373 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1774334 | NM_138820.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1774334 | BC007502.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1774334 | NM_138820.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1774334 | BC007502.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1774334 | NM_138820.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1774334 | BC007502.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1774334 | NM_138820.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1774334 | BC007502.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1774334 | NM_138820.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1774334 | BC007502.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_20270388-S | NM_138820.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_20270388-S | BC007502.2 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_11040-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0004610184 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0004610184 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1774334 | NM_138820.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1774334 | BC007502.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1774334 | NM_138820.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1774334 | BC007502.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 4890398 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_11040 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1774334 | NM_138820.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1774334 | BC007502.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1774334 | NM_138820.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1774334 | BC007502.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1774334 | NM_138820.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1774334 | BC007502.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1774334 | NM_138820.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1774334 | BC007502.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1774334 | NM_138820.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1774334 | BC007502.2 | human_transcriptome |