| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012358.hg.1 |
ENST00000399363.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012358.hg.1 |
ENST00000399363.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012358.hg.1 |
ENST00000399363.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012358.hg.1 |
ENST00000399363.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012358.hg.1 |
ENST00000399363.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012358.hg.1 |
NM_001310219.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012358.hg.1 |
NM_001310219.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012358.hg.1 |
NM_001310219.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012358.hg.1 |
NM_001310219.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012358.hg.1 |
NM_001310219.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700205341.hg.1 |
ENST00000399363.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700205341.hg.1 |
NM_001310219.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700205341.hg.1 |
ENST00000399363.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700205341.hg.1 |
NM_001310219.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700205341.hg.1 |
TC1700012358.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700012358.hg.1 |
ENST00000399363.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012358.hg.1 |
ENST00000399363.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012358.hg.1 |
ENST00000399363.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012358.hg.1 |
ENST00000399363.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012358.hg.1 |
ENST00000399363.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700012358.hg.1 |
NM_001310219.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012358.hg.1 |
NM_001310219.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012358.hg.1 |
NM_001310219.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012358.hg.1 |
NM_001310219.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700012358.hg.1 |
NM_001310219.1 |
Refseq |