| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006462.hg.1 |
ENST00000358105.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006462.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006462.hg.1 |
ENST00000512931.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006462.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006462.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006462.hg.1 |
TC04002937.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000502497.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000502497.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000502497.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000502497.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000502497.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000503883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000503883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000503883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000503883.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000508314.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000508314.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000508314.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000621014.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
ENST00000621014.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006464.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006464.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006464.hg.1 |
TC04002937.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000502497.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000503883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000502497.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000503883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188583.hg.1 |
TC0400012821.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000358105.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000502497.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000502497.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000502497.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000502497.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000502497.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000503883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000503883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000503883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000503883.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000508314.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000508314.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000508314.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000512931.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000513920.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000514249.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000621014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
ENST00000621014.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012821.hg.1 |
NM_152770.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188585.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188585.hg.1 |
NM_001206997.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400188585.hg.1 |
ENST00000508675.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400188585.hg.1 |
TC0400012821.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400188585.hg.1 |
NM_001206997.1 |
Refseq |