| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000555101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000555101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000555101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000556788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000556788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000557175.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000557175.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000557175.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000557175.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
ENST00000557175.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013381.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013381.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013381.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14013381.hg.1 |
TC14002322.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000555101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000555101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000555101.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000556788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000556788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000557175.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000557175.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000557175.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000557175.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
ENST00000557175.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010754.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400182485.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182485.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182485.hg.1 |
ENST00000557175.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182485.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400182485.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182485.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182485.hg.1 |
ENST00000557175.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182485.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400182485.hg.1 |
TC1400010754.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400182486.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182486.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182486.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400182486.hg.1 |
ENST00000267485.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182486.hg.1 |
ENST00000334342.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400182486.hg.1 |
NM_001001872.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400182486.hg.1 |
TC1400010754.hg.1 |
Clariom D TC |