| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15008509.hg.1 |
ENST00000333334.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15008509.hg.1 |
NM_153040.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15008509.hg.1 |
TC15000885.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
ENST00000556865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
ENST00000556865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
ENST00000556865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
ENST00000556865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
ENST00000624458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
ENST00000624458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
ENST00000624458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
ENST00000624458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
ENST00000624458.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
NM_153040.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
NM_153040.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
NM_153040.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
NM_153040.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500008374.hg.1 |
NM_153040.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500160721.hg.1 |
ENST00000556865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500160721.hg.1 |
ENST00000624458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500160721.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500160721.hg.1 |
NM_153040.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500160721.hg.1 |
TC1500008374.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500160721.hg.1 |
NM_153040.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500160721.hg.1 |
ENST00000556865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500160721.hg.1 |
ENST00000624458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500160721.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
ENST00000556865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
ENST00000556865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
ENST00000556865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
ENST00000556865.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
ENST00000624458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
ENST00000624458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
ENST00000624458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
ENST00000624458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
ENST00000624458.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
NM_001301106.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
NM_153040.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
NM_153040.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
NM_153040.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
NM_153040.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500008374.hg.1 |
NM_153040.2 |
Refseq |